Value Plastics Luer Fittings and Adapters
Supplier: Avantor Fluid Handling
15973-45BLK
MFLB15973-45BLKEA 2189.6 SGD
MFLB15973-45BLK
Value Plastics Luer Fittings and Adapters
Fittings and Connectors
Premium luer fittings for critical applications.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Manufactured from 100% virgin raw materials in a Class 100000 environment
  • Full material traceability on all items
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Manufactured in compliance with ISO 594-1 standards, these fittigns are inspected to be free from contaminants, and are fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity. ChooseMore Product Information