LEVIFLOW® Single-Use Ultrasonic Flowmeter Starter Kit
Supplier: Avantor Fluid Handling
24500-07
MFLB24500-07EA 0 SGD
MFLB24500-07
LEVIFLOW® Single-Use Ultrasonic Flowmeter Starter Kit
Fluid Sensors
Obtain flow measurements of high-purity fluids.
  • Order the starter kit and a flow sensor to build a complete system
  • Choose from sterile or non-sterile flow sensors covering a range of 0.08 to 800 L/min
  • Avoid contamination— systems don't contain moving or obtrusive parts
  • Order starter bundle for immediate use
These ultrasonic flowmeter systems are designed for use in applications were cleanliness is required. Readings are accurate to 1%—high precision for high-purity fluids. Each system requires a starter kit and clamp-on flow sensor; order separately.More Product Information