CPC® Metal Quick-Disconnect Fittings, Compression Elbow Inserts
Supplier: Avantor Fluid Handling
31309-26BLK
MFLB31309-26BLKEA 0 SGD
MFLB31309-26BLK
CPC® Metal Quick-Disconnect Fittings, Compression Elbow Inserts
Fittings and Connectors
Convenience of a quick-disconnect in pressure and vacuum applications.
  • Constructed of durable chrome-plated brass
  • Easy one-hand connection and disconnection
These CPC® metal quick-disconnect fittings are ideal for pressure and vacuum applications. Fittings are made of chrome-plated brass with stainless steel springs and latch. Valved fittings have an acetal shutoff valve that seals the flow path when fitting halves are disconnected. Can withstand vacuum to 250 psi, temperatures from -40 to 180 °F, and can be used with gases or liquids.
A completeMore Product Information