Grooving Tool for PFA Compression Fittings
Supplier: Avantor Fluid Handling
31321-49
MFLB31321-49EA 800 SGD
MFLB31321-49
Grooving Tool for PFA Compression Fittings
Puller and Inserter Tools
Get a secure seal at higher pressures.
  • Optimize performance of your PFA Compression Fittings
Improve the sealing capability of PTFE fittings. Fittings withstand pressures to 150 psi when a tool is used (40 psi without tool).