Hedland Doppler Ultrasonic Flowmeters
Supplier: Avantor Fluid Handling
32615-80
MFLB32615-80EA 0 SGD
MFLB32615-80
Hedland Doppler Ultrasonic Flowmeters
Flow Meters
In-field linearization allows adaptation to a wide variety of applications.
  • Accuracy is ±2% full-scale over calibrated span
  • Nonintrusive, clamp-on transducers fit pipes from ¼” and above
  • Clamp-on sensor eliminates the need for in-line flanges, pipe fittings, strainers, or filters
Use these doppler flowmeters measure the flow of liquids in pipes from ¼” and larger. The transducers send flow data via RG59 coax cable to the monitor, and the monitor’s software automatically adjusts filtering parameters and gain settings to ensure reliable readings areMore Product Information