Aalborg® Mass Flow Control Command Modules
Supplier: Avantor Fluid Handling
32681-22
MFLB32681-22EA 0 SGD
MFLB32681-22
Aalborg® Mass Flow Control Command Modules
Flow Controllers
Modular system works with any mass flow meter or controller*.
  • Configure and direct the operation of up to 4 controllers either locally, via an RS-232 interface, or via the Internet
  • Use the integrated batch, totalizer, or timer functions with up to 8 relays to control external system devices or alarms
  • Controllers feature exceptional repeatability that reduces quality deviations in any process that relies on multiple devices
This design has a well-established standing in the control industry because of the flexibility and features that are inherent inMore Product Information