AW-Lake Gear Flow Sensor Flow Monitor Transmitters
Supplier: Avantor Fluid Handling
32928-76
MFLB32928-76EA 0 SGD
MFLB32928-76
AW-Lake Gear Flow Sensor Flow Monitor Transmitters
Flow Controllers
Easily control flow of your gear flow sensors.
  • Monitor both flow rate and total flow
Monitor both flow rate and flow from pulse output flowmeters in any engineering unit with a push of a button. Programmable, compact, and easy-to-use. Input square wave, sine, or sawtooth frequency. Back-up totalizer lock-out feature prevents accidental erasure. High-speed microprocessor ensures fast and accurate reporting — includes a 4 to 20 mA output for recording or datalogging. Model 32928-76 has two channels to monitor dual flows and display them separately, as aMore Product Information