Cadence Luer Adapter Fittings, Plated Brass
Supplier: Avantor Fluid Handling
41507-51 41507-52 41507-55 41507-56 41507-57 41507-60 41507-61 41507-62 41507-63 41507-65 41507-66 41507-70 41507-71 41507-72 41507-73 41507-84 41507-85 41507-86 41507-87 41507-54
MFLA41507-53EA 0 SGD
MFLA41507-53 MFLB41507-51 MFLB41507-52 MFLB41507-55 MFLB41507-56 MFLB41507-57 MFLB41507-60 MFLB41507-61 MFLB41507-62 MFLB41507-63 MFLB41507-65 MFLB41507-66 MFLB41507-70 MFLB41507-71 MFLB41507-72 MFLB41507-73 MFLB41507-84 MFLB41507-85 MFLB41507-86 MFLB41507-87 MFLB41507-54
Cadence Luer Adapter Fittings, Plated Brass
Fittings and Connectors
Excellent durability.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Manufactured in an ISO 14
More Product Information