Kodiak® Level Switches, 316 Stainless Steel
Supplier: Avantor Fluid Handling
68964-29
MFLB68964-29EA 396.06 SGD
MFLB68964-29
Kodiak® Level Switches, 316 Stainless Steel
Level Sensors
Accurate and rugged level monitoring at higher temperatures.
  • Constructed of durable 316 stainless steel (SS)
These all-stainless-steel units are exceptionally accurate and rugged for level monitoring at higher temperatures...to 300 °F (GM424113)... and in pressurized or corrosive liquids. Widely used in chemical plants and systems. For use in oils, water, chemicals.