Masterflex® Mechanically Coupled Centrifugal Pumps, Avantor®
Supplier: Avantor Fluid Handling
70725-00 70725-07 70725-10 70725-01 70725-02 70725-06 70725-14
MFLB70725-00EA 0 SGD
MFLB70725-00 MFLB70725-07 MFLB70725-10 MFLB70725-01 MFLB70725-02 MFLB70725-06 MFLB70725-14
Masterflex® Mechanically Coupled Centrifugal Pumps, Avantor®
Pumps Centrifugal Pumps
Chemical-resistance pump heads provide dependable performance and continuous service.
  • Mechanically close-coupled design saves space and simplifies installation and maintenance
  • Back pullout construction — assembly and overhaul of impeller and seal without distorting suction and discharge connections
  • Top centerline discharge and foot support under casing ensure self-venting and reduce misalignment from pipe loads
The mechanically coupled centrifugal pumps, constructed of 304 stainless steel, are ideal for food and pharmaceutical processing plants, asMore Product Information