Masterflex® Mechanically Coupled High-Head Centrifugal Pumps, Avantor®
Supplier: Avantor Fluid Handling
70725-13 70725-11 70725-20 70725-22 70725-26 70725-31 70725-32 70725-36 70725-37
MFLB70725-13EA 0 SGD
MFLB70725-13 MFLB70725-11 MFLB70725-20 MFLB70725-22 MFLB70725-26 MFLB70725-31 MFLB70725-32 MFLB70725-36 MFLB70725-37
Masterflex® Mechanically Coupled High-Head Centrifugal Pumps, Avantor®
Pumps Centrifugal Pumps
Rugged continuous-duty service with chemical and corrosion resistance.
  • Close-coupled design saves space; simplifies maintenance and installation
  • Stainless steel liquid end components deliver corrosion resistance
  • Versatile mounting can be installed horizontally or vertically
  • Back-pullout construction assembly allows overhaul of impeller and seal without disturbing suction and discharge connections
  • Top centerline discharge and foot support under casing ensures self-venting and reduces misalignment from pipe loads
  • High operating efficiency lowers operating
More Product Information