Fluoroplastic Enclosed Motor Chemical-Resistance Centrifugal Pumps
Supplier: Avantor Fluid Handling
72011-00 72011-05 72011-10
MFLB72011-00EA 0 SGD
MFLB72011-00 MFLB72011-05 MFLB72011-10
Fluoroplastic Enclosed Motor Chemical-Resistance Centrifugal Pumps
Pumps Centrifugal Pumps
Twin bearing system handles thrust loads for extended service life.
  • Quiet, energy efficient, and compact
  • Internal cooling circuit reduces bearing temperature
  • Enclosed motor for use in harsh environments
Fluoroplastic pumps with carbon fiber-reinforced ETFE are designed for highly corrosive or high-purity applications.
Delivery information: Includes a 6-ft (1.8 m) cord with stripped ends.