Masterflex® Microscope Slides with Wells, Avantor®
Supplier: Avantor Fluid Handling
75955-27
MFLB75955-27EA 0 SGD
MFLB75955-27
Masterflex® Microscope Slides with Wells, Avantor®
Slides Microscope Slides
Keep samples isolated.
  • Screen-printed well design prevents bleed-through
  • Hydrophobic epoxy ink withstands frequent washing and resists chemicals
  • Contrasting ink makes orienting the microscope field of view over a sample easy
These printed microscope slides feature circular wells designed to hold samples in place, allowing multiple wells per slide, so you can increase throughput without adding to cleanup. Black PTFE-based epoxy makes locating wells with a microscope simple and quick, improving efficiency. Select from a range of well counts to suit any application.