Masterflex® Air-Operated Drum Pumps, Avantor®
Supplier: Avantor Fluid Handling
79015-00
MFLB79015-00EA 0 SGD
MFLB79015-00
Masterflex® Air-Operated Drum Pumps, Avantor®
Pumps Drum/Transfer Pumps
Extremely low noise level for busy work environments due to unique air mechanism.
  • 60% lower air consumption
  • Quick an deasy maintenance with on piece air mechanism
  • Pumps provide fast, reliable delivery and are rugged enough for extreme conditions
  • Use for any application requiring pressurized oil — ideal for manufacturing, construction, and mining
Pumps handle petroleum-based lubricants and synthetic oils. They also feature an internal muffler to reduce air noise. Stall-free air motor can be used with or without a lubricator. Make quick orientationMore Product Information