Edwards Vacuum Solenoid Right-Angle Valves
Supplier: Avantor Fluid Handling
79302-78
MFLB79302-78EA 0 SGD
MFLB79302-78
Edwards Vacuum Solenoid Right-Angle Valves
Valves Solenoid Valves
Designed to maintain vacuum levels in your systems.
  • Easily control vacuum levels with these solenoid valves
Compact and lightweight valves offer a pressure range from 8 x 10⁻¹⁰ to 150 Torr.