Electrically and Pneumatically Actuated Elliptical Valves
Supplier: Avantor Fluid Handling
98153-16 98154-13 98153-03 98153-15 98153-30 98153-33
MFLB98153-16EA 0 SGD
MFLB98153-16 MFLB98154-13 MFLB98153-03 MFLB98153-15 MFLB98153-30 MFLB98153-33
Electrically and Pneumatically Actuated Elliptical Valves
Valves Elliptical Valves
Completely disassemble in-line for easy cleaning.
  • FDA-compliant
  • Choose between Polypropylene (PP) or PVDF valve options
Use these actuated elliptical valves for liquid, gas, and vacuum applications. Valve systems are available in PP or PVDF bodies; all are FDA compliant and have O-rings made of Viton®. Rated for 150 psi at 73 °F (10 bar at 22 °C) (liquids).