Chemically Inert Two-Way Ball Valves
Supplier: Avantor Fluid Handling
98402-10
MFLB98402-10EA 0 SGD
MFLB98402-10
Chemically Inert Two-Way Ball Valves
Valves Ball Valves
Safely control flow in chemically aggressive applications.
  • Constructed of chemically inert PFA
All wetted materials are PFA—ideal for use with corrosive chemicals. Simply make a 1/4-turn to start or stop the flow. Maximum pressure is 150 psi; (10.3 bar) fluid temperature range is 70 to 400 °F (21 to 204 °C); operating temperature range is 0 to 150 °F (–18 to 66 °C).