Two-Way Ball Valves with Lockout/Tagout Assembly
Supplier: Avantor Fluid Handling
98711-04
MFLB98711-04EA 0 SGD
MFLB98711-04
Two-Way Ball Valves with Lockout/Tagout Assembly
Valves Ball Valves
Provides security and safety where most needed.
  • Smooth flow path reduces pressure loss
  • Corrosion-resistant thermoplastic ensures long life
  • PTFE shaft bearing and seats eliminate friction and wear
These lockout/tagout ball valves ensure safety and control in your process lines. The design accepts most OSHA approved locks (not included). Lockout/tagout assembly is designed with an upper and lower hasp and is easily retrofitted in the field to lock in either open or closed position. Available in PVC or CPVC for higher temperature applications. All valves feature FKM seals.