BucketBinders™ Multi Bucket Systems
Supplier: MICRONOVA
BB-DBL-DBYE
MICRBB-DBL-DBYEEA 0 SGD
MICRBB-DBL-DBYE
BucketBinders™ Multi Bucket Systems
Mops
BucketBinders™ bucket system is a cost effective, ergonomic and simple that replaces the need for bulky carts.
  • Easy clip action allows attachment before or after bucket is filled with disinfectant
  • Polyketone hooks firmly secure two or three rectangular buckets together
  • Stainless steel base plate attaches at caster for support
  • Sturdy enough to secure two or three buckets and easy to maneuver with up to 15 litres of liquid in each bucket
  • Lightweight; less than 1 kg per pair
  • Allows scale up or down as required
  • Autoclavable
The system offers easy movement,More Product Information