Cleanroom pens
Supplier: MICRONOVA
PEN-01IR PEN-02IR PEN-20IR PEN-02 PEN-01TIR PEN-25IR PEN-22IR PEN-51IR
MICRPEN-01IREA 0 SGD
MICRPEN-01IR MICRPEN-02IR MICRPEN-20IR MICRPEN-02 MICRPEN-01TIR MICRPEN-25IR MICRPEN-22IR MICRPEN-51IR
Cleanroom pens
Pens Cleanroom Pens
Micronova cleanroom pens feature low sodium, indelible ink and a special low particulate barrel to limit particle generation. Cleanroom irradiated Sharpie® markers write permanently on most surfaces with quick drying, non toxic, alcohol-based ink that is fade and water resistant to further extend multiple project uses. Marks will not smear on reusable glass, metal, or porcelain lab ware, but can be scrubbed off.
  • Indelible, low sodium ink pens
  • White, high density polypropylene barrel
  • Holds up to wipe down with IPA, bleach, and phenols; integrity of printed
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable