Flathead mop covers and hardware, Slimline™
Supplier: MICRONOVA
SLSC6-9 SLSC6-14 SLSC6-18 SLSC6-14IR SLSC6AA-14 SLSC6-9IR SLSC6-18IR
MICRSLSC6-9EA 0 SGD
MICRSLSC6-9 MICRSLSC6-14 MICRSLSC6-18 MICRSLSC6-14IR MICRSLSC6AA-14 MICRSLSC6-9IR MICRSLSC6-18IR
Flathead mop covers and hardware, Slimline™
Mops
A series of elasticised slipcovers, with urethane foam interiors, that are ideal for fine cleaning and sanitising all surfaces and equipment in the cleanroom. Available without a foam interior or gamma irradiated for aseptic environments.
  • Covers are easy to change and fit securely over mop frame
  • Fully encased Urethane Foam layer for ample disinfectant dispersal
  • Single-ply covers available without foam interior
  • Fits on QPSL plastic frame or QDSL stainless steel frame
  • Suitable for use with strong disinfectants and repeated autoclaving
The MicronovaMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Waste Reducing