Wipes, pre-saturated with silicone, SilSat™
Supplier: MICRONOVA
SSW6-01IR
MICRSSW6-01IREA 0 SGD
MICRSSW6-01IR
Wipes, pre-saturated with silicone, SilSat™
Wipes Disposable Wipes and Towels Presaturated Wipes
SilSat wipes address the problem of using lubricant in a controlled application by offering high grade silicone on a pre-saturated wipe. Each polyester wipe carries several grams of silicone, enough to service gears or rollers on fill lines or lubricate stopper caps and valves. Wipes are gamma irradiated.
  • High grade silicone/alcohol blend
  • Light impregnation
  • Avoids possible transfer of silicon into product or other critical surfaces
Colour: White
Weight: 106 g/m²