Laboratory washer-Disinfector, PG 8527
Supplier: MIELE
6881690 6881680
MIEL6881690EA 0 SGD
MIEL6881690 MIEL6881680
Laboratory washer-Disinfector, PG 8527
Glassware Washing Machines
The washer-disinfector PG 8527 offers great added value for central and decentralised processing of large volumes of instruments: greater cleaning capacity, greater process reliability and greater economy.
  • New Profitronic controller with PerfectTouch control
  • Patented maintenance-free conductivity meter
  • Improved hygienic wash chamber with a unique spray arm monitoring system
  • Highly accurate detergent dispensing monitor using ultrasound technology
  • Improved HEPA-filtered forced air drying system
  • Full-view glass door with chamber lighting
Construction:More Product Information