Transfer trolley
Supplier: MIELE
7397640
MIEL7397640EA 0 SGD
MIEL7397640
Transfer trolley
Trolleys
For loading and unloading washers with backets and inserts.
  • With removable drip tray (W×D×H: 603×866×70 mm)
  • Both ends can dock onto machine, tables, sluices or conveyors
  • Docking height 850, –100, + 150 mm
  • 4 lockable wheels, Ø 100 mm
Order Now
SPECIFICATIONS
Environmentally Preferable