Bags, zip seal, anti-UV, writing area, Minigrip®
Supplier: Flexico
ITM025704
MINIITM025704EA 0 SGD
MINIITM025704
Bags, zip seal, anti-UV, writing area, Minigrip®
Bags Sample Bags
PE bags, thickness 80 µm.
  • UV protection
  • 3 white write-on panels
  • Resealable, reusable, 100% recyclable
Made of polyethylene (LDPE). UV protection provides barrier properties for the content of the bag (wavelengths between 280 and 350 nm) and can withstand an irradiation energy of 200 Kly. Zip seal designed for easy opening and high speed packing. The Minigrip® bag provides an excellent mechanical resistance and high temperature resistance (from −40°C up to +60°C). NOT suitable for food contact.