Multi-purpose trolleys
Supplier: MULTIROIR CONTROLEC
65005HS808E3 65003HS808LE 65001HS808LE3 65002HS808LA3
MULC65005HS808E3EA 0 SGD
MULC65005HS808E3 MULC65003HS808LE MULC65001HS808LE3 MULC65002HS808LA3
Multi-purpose trolleys
Trolleys
These multi-use trolleys have been designed for intensive use. They are made of an aluminium frame with heavy duty polypropylene trays.
  • Grey or black
  • 2 or 3 tray versions
  • Unequalled load capacity: 80 kg per tray and 250 kg per trolley
  • 4 ball bearing wheels diam. 100 mm, 2 with brakes
The trolleys are light, easy to handle and very robust.