Muffle furnaces with fold down door, Tmax 1100 °C and controller R7, LE series (LE 1/11 - LE24/11)
Supplier: NABERTHERM
LE062K17N1
NABELE062K17N1EA 7650 SGD
NABELE062K17N1
Muffle furnaces with fold down door, Tmax 1100 °C and controller R7, LE series (LE 1/11 - LE24/11)
Ovens Furnaces (Ashing and Muffle Ovens)
With their convincing price/performance ratio and the fast heat-up rates, these compact muffle furnaces are perfect for many applications in the laboratory. Quality features like the dual-shell furnace housing of rust-free stainless steel and their compact, lightweight construction. The heating elements encased in quartz glass tubes make these models reliable partners for your application. The furnaces have Tmax 1100 °C, working temperature of 1050 °C.
  • Heating from two sides via heating elements protected in quartz glass tubes
  • Fast heating times
  • Exclusive
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable