Benchtop Forced Convection Chamber Furnaces, with Tmax 650 or 850 °C
Supplier: NABERTHERM
NAT-0161L1
NABENAT-0161L1EA 0 SGD
NABENAT-0161L1
Benchtop Forced Convection Chamber Furnaces, with Tmax 650 or 850 °C
Ovens Furnaces (Ashing and Muffle Ovens)
These forced convection chamber furnaces provide good temperature uniformity. Due to their compact benchtop design, this series is very well suited for use in laboratories or areas with limited space.
  • Choice of models with Tmax 650 or 850 °C
  • Horizontal air circulation with optimum distribution via stainless steel baffles
  • Dual shell housing made of textured stainless steel sheets with additional fan cooling for low surface temperature
  • Integrated control unit
  • Swing door is hinged on the right side, door opening temperatures up to 400 °C
  • Optimum air
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable