Compact Tube Furnaces R and RD Series
Supplier: NABERTHERM
RD30A171A
NABERD30A171AEA 0 SGD
NABERD30A171A
Compact Tube Furnaces R and RD Series
Ovens Tube Furnaces
These compact bench top tube furnaces with integrated controllers can be widely used for a variety of processes. Equipped with a standard C 530 ceramic working tube and two fibre plugs they have an unbeatable price/performance ratio. Units feature rust-free, textured stainless steel exteriors. R models feature Tmax 1200 or 1300 °C, RD models have Tmax 1100 or 1300 °C.
  • Tube diameter: RD models 30 mm, R models 50 to 170 mm
  • Heated length: RD models 200 mm, R models 250 to 1000 mm
  • C 530 ceramic working tube and two fibre plugs supplied as standard
  • R and RD
More Product Information