Acrylic Imhoff cone rack, Nalgene®
Supplier: Thermo Fisher Scientific
1001-0010
NALG1001-0010EA 262 SGD
NALG1001-0010
Acrylic Imhoff cone rack, Nalgene®
Retort Stands
Securely hold three Imhoff cones with the durable and transparent Thermo Scientific™ Nalgene™ Acrylic Imhoff Cone Rack, featuring a wide base for stability on the lab bench.
  • Transparent; easily see level of contents
  • Can also be used to hold 500 and 1000 ml Nalgene separatory funnels