Bottle top vacuum filtration systems, cellulose nitrate membrane, Nalgene®
Supplier: Thermo Fisher Scientific
140-4045
NALG140-4045EA 0 SGD
NALG140-4045
Bottle top vacuum filtration systems, cellulose nitrate membrane, Nalgene®
Filters Bottle Top Filters
These filters are particularly useful for large volume microbiological QC samples.
  • Convenient snap-away design
  • Compatible with filter manifolds and filtering flasks
Certifications: Membrane conforms to APHA standard methods, 16th edition, section 909A, B, C. Meets EPA requirements for water quality work.
Delivery information: Upper chamber with reference mark at 150 ml; PS cover; CN membrane; support pad; PS support plate/funnel that fits a stopper or adapter with ⁹/₁₆" hole; quick-disconnect adapter for ¹/₄ or ⁵/₁₆" tubing. Each pack of 12 funnelsMore Product Information