Bottles, narrow neck, round, FEP, with screw caps, Nalgene®
Supplier: Thermo Fisher Scientific
1600-0004 1600-0008 1600-0016 1600-0032 1600-0064 1600-0002 1600-0001
NALG1600-0004EA 0 SGD
NALG1600-0004 NALG1600-0008 NALG1600-0016 NALG1600-0032 NALG1600-0064 NALG1600-0002 NALG1600-0001
Bottles, narrow neck, round, FEP, with screw caps, Nalgene®
Bottles Plastic Bottles
FEP, translucent, with ETFE screw cap.
  • Highest chemical resistance
  • Temperature resistant from −105 to +150 °C, leakproof closure
  • Autoclavable
Bottles with the highest chemical and corrosion resistance. Suitable for applications at low and high temperatures, trace analyses, and for many types of work with organic solvents. Suitable for use in cleanrooms, analysis and for storage.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification NAR Certificate of Retirement