Carboys, wide neck, HDPE, heavy duty, Nalgene®
Supplier: Thermo Fisher Scientific
2214-0050
NALG2214-0050EA 0 SGD
NALG2214-0050
Carboys, wide neck, HDPE, heavy duty, Nalgene®
Bottles Carboys
HDPE, translucent with HDPE closure.
  • Good chemical resistance
  • Heat resistant from -100 to +120 °C
  • Robust, leakproof closure, meets the requirements of regulation 177.1520 governing storage of foodstuffs
For safe storage and shipping of chemicals. Protects substances that must not lose their consistency and must satisfy specifications for low levels of particle contamination. The jerrycan with wide opening (70 mm Ø) has been constructed as a system with leak-proof sealing. The wide mouth aperture makes filling, emptying and cleaning easy. A practicalMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification NAR Certificate of Retirement