Venting filters for carboys, Nalgene®
Supplier: Thermo Fisher Scientific
223-0030
NALG223-0030EA 0 SGD
NALG223-0030
Venting filters for carboys, Nalgene®
Filters Venting Filters
These vent filters with hydrophobic PTFE membrane and PP housing are designed to maintain purified water stored in carboys. They can also be used for venting and solvent filtration applications.
  • Use up to five times in venting applications
  • 6 ml liquid hold-up volume