Carboys, with sanitary flanges, PC, Nalgene®
Supplier: Thermo Fisher Scientific
2261-0050
NALG2261-0050EA 0 SGD
NALG2261-0050
Carboys, with sanitary flanges, PC, Nalgene®
Bottles Carboys
PC, clear.
  • Good chemical resistance
  • Thermally resistant from -100 to +135 °C, autoclavable
  • Material complies with USP class VI and FDA specifications
Ideal for storing, transporting or dispensing in pharmaceutical and biotechnology applications. The welded 75 mm sanitary flange fits standard flange connectors. The carboy is sealed with a fixed, securely fitted clamp fastener. The sanitary design is easier to clean than threaded containers.