Dollys, Nalgene®
Supplier: Thermo Fisher Scientific
2624-0020
NALG2624-0020EA 0 SGD
NALG2624-0020
Dollys, Nalgene®
Trolleys
For transporting round tanks, carboys and balloon flasks as well as other round containers with up to 521 mm Ø.
  • Resistant to corrosion and to acids and alkalis
  • Strong, maximum load capacity 227,3 kg
  • Casters designed to minimise stress on flooring
Stainless steel, rustproof.
Order Now
SPECIFICATIONS
Environmentally Preferable