Nalgene™ Carboys, Narrow Neck, Single Use, HDPE
Supplier: Thermo Fisher Scientific
342289-0050
NALG342289-0050EA 0 SGD
NALG342289-0050
Nalgene™ Carboys, Narrow Neck, Single Use, HDPE
Bottles Carboys
HDPE, translucent with white PP closure.
  • Good chemical resistance
  • Heat resistant from –100 to +120 °C
  • Complies with the requirements of USP class VI, non cytotoxic
Safer than glass. Ideal for storing sterile liquids and pharmaceutical/biotechnology products. The extra wide mouth makes filling and emptying easier.
Eliminate cleaning, prep and set-up costs with ready-to-use Nalgene™ single-use HDPE carboys. Supplied sterile for one-time use, the high-density polyethelyne vessels are ideal for storing sterile fluids and pharmaceutical/biotech products.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification NAR Certificate of Retirement