Racks for cryogenic vials, Nalgene®
Supplier: Thermo Fisher Scientific
5030-0505
NALG5030-0505EA 0 SGD
NALG5030-0505
Racks for cryogenic vials, Nalgene®
Racks Cryogenic Vial Racks
PC, autoclavable.
  • Bottom interlocks with base of vial
  • Spacing around each hole allows vials to be easily removed
Allows single handed vial manipulation.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification IRECS; ISO 14001 (Certificate Registration No. EMS 752603)