Rack for cryogenic vials, Nalgene®
Supplier: Thermo Fisher Scientific
5030-0510
NALG5030-0510EA 0 SGD
NALG5030-0510
Rack for cryogenic vials, Nalgene®
Racks Cryogenic Vial Racks
PC, autoclavable.
  • For 5×10 cryovials outer-Ø 13,5 mm
Allows single handed vial manipulation, bottom of each well interlocks with base of vial.
W×D×H: 197×102×28 mm
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification IRECS; ISO 14001 (Certificate Registration No. EMS 752603)