Racks for cryoboxes, Nalgene®
Supplier: Thermo Fisher Scientific
5036-0004 5036-0009 5038-4322 5038-4422 DS5035-0004 DS5035-0009 DS5037-0004 DS5037-0007
NALG5036-0004EA 0 SGD
NALG5036-0004 NALG5036-0009 NALG5038-4322 NALG5038-4422 NALGDS5035-0004 NALGDS5035-0009 NALGDS5037-0004 NALGDS5037-0007
Racks for cryoboxes, Nalgene®
Racks Cryogenic Box Racks
Stainless steel.
Retainers hold each box securely but separately and allow fast retrieval of one box and immediate return to rack.
Vertical
479-3351, 479-3352: for cryoboxes with 5×5 cryotubes
212-8117, 212-8118: for standard cryoboxes
479-3356, 479-3357: for cryoboxes with 5 ml tubes and Mr. Frosty
Horizontal
Fit most upright freezer compartments
For standard cryoboxes of 133×133×51 mm
With two lateral handles
Delivery information: Supplied without cryoboxes.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification NAR Certificate of Retirement