Desiccator, Nalgene®
Supplier: Thermo Fisher Scientific
5311-0250
NALG5311-0250EA 252 SGD
NALG5311-0250
Desiccator, Nalgene®
Desiccators Desiccators, Plastic
Transparent PC, unbreakable and lightweight. Suitable for vacuum and non vacuum applications. Has wide base for greater stability. Accepts 6,4 mm (¹/₄") to 19 mm (³/₄") int.Ø. vacuum tubing and can be used with any standard 230 mm desiccator plate, e.g. 467-3100. Should be used at room temperature only.
  • Convenient side arms on lid and base
  • O-ring ensures an airtight seal, and does not require greasing
  • Removable stopcock
Ordering information: Supplied without plate, please order suitable plate separately.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification IRECS; ISO 14001 (Certificate Registration No. EMS 752603)