Desiccator, Nalgene®
Supplier: Thermo Fisher Scientific
5315-0150
NALG5315-0150EA 0 SGD
NALG5315-0150
Desiccator, Nalgene®
Desiccators Desiccators, Plastic
Ideal for drying and drying processes. Cover is transparent, robust PC with recessed rim. Base is durable blue PP.
  • Base is impervious to all solid desiccants
  • Autoclavable
Delivery information: Supplied with 140 mm aluminium plate.