Bottle top vacuum filtration systems, PES membrane, Rapid-Flow™ Nalgene®
Supplier: Thermo Fisher Scientific
564-0020
NALG564-0020EA 191 SGD
NALG564-0020
Bottle top vacuum filtration systems, PES membrane, Rapid-Flow™ Nalgene®
Filters Bottle Top Filters
Rapid-Flow™ disposable filter units with PES membrane, PS funnel and PP collection tube feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination and optimal choice for small volume tissue culture work. Offers clean, fast filtration of 20 to 50 ml of media or buffers.
  • Color coded according to membrane
  • Fastest flow rate and lowest clogging of comparable alternatives
  • 0.1 µm PES available for mycoplasma protection
  • 0.2 µm PES devices are stem cell and
More Product Information