Petri dish racks, Nalgene®
Supplier: Thermo Fisher Scientific
5920-0060
NALG5920-0060EA 262 SGD
NALG5920-0060
Petri dish racks, Nalgene®
Racks Petri Dish Racks
Petri dish rack with PC frame and white PC posts.
  • Smooth, rounded corners
  • Finger-grip handle and rubber feet make carrying safe and secure
  • Rack can be decontaminated by heat sterilisation
For up to 54 plastic round dishes (100×15 mm) or 42 glass round dishes (100×15 mm). Simple assembly required.
W×D×H: 203×343×235 mm