Vacuum check valve and positive connector, Nalgene®
Supplier: Thermo Fisher Scientific
6120-0010
NALG6120-0010EA 0 SGD
NALG6120-0010
Vacuum check valve and positive connector, Nalgene®
Valves Non Return Valves
These vacuum check valves with positive connector are designed to prevent back pressure with aspirator-type vacuum pumps.
  • Work best between 10- and 28- in. Hg.
  • Silicone diaphragm prevents back pressure
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification NAR Certificate of Retirement