Vacuum aspirator pump, Nalgene®
Supplier: Thermo Fisher Scientific
6140-0010
NALG6140-0010EA 0 SGD
NALG6140-0010
Vacuum aspirator pump, Nalgene®
Pumps Vacuum Aspiration Systems
Attach the Nalgene™ aspirator vacuum pump directly to sink faucets for vacuum draw. It can draw 11,5 L air per minute at a water flow rate of 6,5 L per minute. It operates efficiently with water pressures as low as 7,5 psig for minimum water consumption and comes with an integral check valve and tubing for insertion in discharge end.
  • Attaches to a standard faucet with ³/₈" NPT connecting thread for easy installation
  • Ultimate achievable vacuum of 724 mm Hg
  • Integral check valve that prevents loss of vacuum when water source is disrupted and prevents backflow
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification NAR Certificate of Retirement