Stirring rods, Nalgene®
Supplier: Thermo Fisher Scientific
6169-0010
NALG6169-0010EA 72.4 SGD
NALG6169-0010
Stirring rods, Nalgene®
Stirrers Stirring Rods
Stirring rod with flattened ends forming flat paddle 16×16 mm at one end and triangular paddle at the other end.
  • Good chemical resistance
  • Autoclavable