Bags, zip seal, Nalgene®
Supplier: Thermo Fisher Scientific
6255-0406 6255-0508 6255-0613 6255-0913 6255-0918
NALG6255-0406EA 0 SGD
NALG6255-0406 NALG6255-0508 NALG6255-0613 NALG6255-0913 NALG6255-0918
Bags, zip seal, Nalgene®
Bags Sample Bags
Ideal for small lab items, botanical samples and other specimens. Not recommended for storing liquids. LDPE, 101 µm.
  • Strong, transparent and waterproof
  • Heavy-duty zipper seals contents
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification NAR Certificate of Retirement