Cryoware marker set, Nalgene®
Supplier: Thermo Fisher Scientific
6313-0010 6313-0020
NALG6313-0010EA 37.2 SGD
NALG6313-0010 NALG6313-0020
Cryoware marker set, Nalgene®
Pens Cryo Markers
For permanent marking of cardboard, cloth labels and plastic.
  • Ink will not fade at ultra-low temperatures
  • Extra fine point
Set consists of 1 red, 1 green, 1 blue and 1 black marker.