Safety bottle carrier with lid, Nalgene®
Supplier: Thermo Fisher Scientific
6505-0010
NALG6505-0010EA 181 SGD
NALG6505-0010
Safety bottle carrier with lid, Nalgene®
Carriers Bottle Carriers/Containers
To hold and carry up to six 500 ml standard reagent bottles, and most other glass or plastic 500 ml bottles, this carrier consists of a bottle basket and a tight fitting cover.
LDPE.
  • Stackable
  • Holds bottles securely
  • Minimise risk of bottles smashing together, falling out or breaking
  • No splashing of dangerous chemicals if glass bottles break
  • Sturdy, wide grip for easy carrying